Assay Details
| Gene Symbol: Hsd17b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010475.1 | NP_034605.1 | 4 - 5 | 569 | 54 | |
| GenBank mRNA | AK019406.1 | - | 1 - 2 | 87 | 54 | |
| AK143346.1 | - | 4 - 5 | 518 | 54 | ||
| BC125657.1 | - | 4 - 5 | 568 | 40130172 | 54 | |
| BC125659.1 | - | 4 - 5 | 568 | 40130174 | 54 | |
| X89627.1 | - | 4 - 5 | 569 | 54 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hsd17b1
Genomic Map