Assay Details
Gene Symbol: Coasy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF521095.1 | - | 6 - 7 | 1337 | 70 | |
AK004878.1 | - | 6 - 7 | 1397 | 70 | ||
AK159405.1 | - | 6 - 7 | 1386 | 70 | ||
AK159912.1 | - | 5 - 6 | 1686 | 70 | ||
AK166894.1 | - | 5 - 6 | 1680 | 70 | ||
AK170330.1 | - | 5 - 6 | 1695 | 70 | ||
BC020046.1 | - | 5 - 6 | 807 | 70 | ||
BC064753.1 | - | 5 - 6 | 1325 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
