Assay Details
Gene Symbol: Coasy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF521095.1 | - | 9 - 10 | 1665 | 75 | |
AK159405.1 | - | 9 - 10 | 1714 | 75 | ||
AK159912.1 | - | 8 - 9 | 2014 | 75 | ||
AK166894.1 | - | 8 - 9 | 2008 | 75 | ||
AK170330.1 | - | 8 - 9 | 2023 | 75 | ||
BC020046.1 | - | 8 - 9 | 1135 | 75 | ||
BC064753.1 | - | 8 - 9 | 1653 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28071820 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28071819 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28071818 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2117843 | AK170330.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2117843 | BC064753.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2117843 | AK159405.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2117843 | AK159912.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2117843 | NM_027896.4 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P136099 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 531857 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 366023 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P136099 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2117843 | AK170330.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2117843 | BC064753.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2117843 | AK159405.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2117843 | AK159912.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2117843 | NM_027896.4 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P101255 | AK170330.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P101255 | BC020046.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P101255 | AK159405.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P101255 | AK159912.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P101255 | AK166894.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28071822 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28071803 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28071836 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28071815 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28071809 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28071817 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28071839 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28071833 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28071850 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28071843 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28071838 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28071805 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28071812 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28071841 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28071824 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28071844 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28071847 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28071825 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28071834 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28071831 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28071821 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28071832 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28071849 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28071851 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28071826 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28071807 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28071829 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28071819 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28071845 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07465577 | - | - |
Whole Mouse Genome G4122A | A_51_P101255 | AK170330.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P101255 | BC020046.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P101255 | AK159405.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P101255 | AK159912.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P101255 | AK166894.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136099 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09529247 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09717054 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09101734 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09101732 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09529246 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09101723 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09529242 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09529240 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09101721 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09529249 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09717032 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09101722 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28071818 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28071820 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28071819 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071921 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071908 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071874 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071764 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071781 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071888 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071893 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071903 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071829 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071786 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071762 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071822 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071891 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071824 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071847 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071760 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071863 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071923 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071797 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071915 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071831 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071809 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071928 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071854 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071807 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071788 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071754 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071833 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071901 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071755 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071895 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071917 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071905 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071884 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071920 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071882 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071793 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071782 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071757 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071856 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071898 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071930 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071817 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071929 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071849 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071869 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071912 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071873 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071900 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071790 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071858 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071791 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071784 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071845 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071841 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071803 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071862 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071815 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071886 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071926 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071819 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071885 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071910 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071843 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071850 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071836 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071838 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071821 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071780 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071826 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071860 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071906 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071806 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071834 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071871 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071825 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071812 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28071867 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09101715 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28071824 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28071841 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09529252 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28071845 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09101725 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09101713 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28071825 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28071809 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28071832 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28071815 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28071821 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28071836 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28071849 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28071838 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28071805 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09101726 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28071817 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28071819 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28071847 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28071834 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28071850 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28071843 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28071803 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09101720 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09529242 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10381234 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10381230 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10381233 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10381231 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10381235 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10381236 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10381229 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10381228 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10381232 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10381227 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10381237 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1423701_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1443829_PM_x_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10381226 | - | - |
Mouse Genome 430 2.0 Array | 1423701_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1423701_at | - | - |
Mouse Genome 430 2.0 Array | 1443829_x_at | - | - |
Mouse Genome 430 2.0 Array | 1443829_PM_x_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71743_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71743_PM_at | - | - |
Mouse Expression Set 430 | 1423701_PM_at | - | - |
Mouse Expression Set 430 | 1423701_at | - | - |
Mouse Expression Set 430 | 1443829_x_at | - | - |
Mouse Expression Set 430 | 1443829_PM_x_at | - | - |
Murine Genome U74v2 Set | 161554_r_at | - | - |
Murine Genome U74v2 Set | 94241_PM_at | - | - |
Murine Genome U74v2 Set | 94241_at | - | - |
Murine Genome U74v2 Set | 161554_PM_r_at | - | - |
Mouse Genome 430A 2.0 Array | 1423701_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1423701_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1423701_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1423701_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1443829_PM_x_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1443829_x_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 6220707 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006220707 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2741236 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2741236 | - | - |
mouse-6 v1.1 (Illumina) | scl071743.9_29-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6220707 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl071743.9_29-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6220707 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2741236 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2741236 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl071743.9_29-S | - | - |
