Assay Details
| Gene Symbol: Slc7a11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011990.2 | NP_036120.1 | 8 - 9 | 1364 | 59 | |
| GenBank mRNA | AB022345.1 | - | 8 - 9 | 1331 | 59 | |
| AK076322.1 | - | 8 - 9 | 1339 | 59 | ||
| AK089202.1 | - | 8 - 9 | 1337 | 59 | ||
| AK162233.1 | - | 8 - 9 | 1139 | 59 | ||
| AK163168.1 | - | 8 - 9 | 1338 | 59 | ||
| AK165599.1 | - | 8 - 9 | 1340 | 59 | ||
| AK171192.1 | - | 8 - 9 | 1362 | 59 | ||
| AK171252.1 | - | 8 - 9 | 1338 | 59 | ||
| AY766236.1 | - | 8 - 9 | 1331 | 59 | ||
| AY766237.1 | - | 1331 | 59 | |||
| BC141402.1 | - | 8 - 9 | 1261 | 9056015 | 59 | |
| BC141408.1 | - | 8 - 9 | 1261 | 9056021 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc7a11
Genomic Map