Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix HT MG-430 PM Array Plate |
1454539_PM_at |
- |
- |
Affymetrix HT MG-430 PM Array Plate |
1433280_PM_at |
- |
- |
Mouse Genome 430 2.0 Array |
1433280_PM_at |
- |
- |
Mouse Genome 430 2.0 Array |
1454539_at |
- |
- |
Mouse Genome 430 2.0 Array |
1433280_at |
- |
- |
Mouse Genome 430 2.0 Array |
1454539_PM_at |
- |
- |
Affymetrix GeneChip Mouse Genome 430 2.0 Array |
71182_at |
- |
- |
Affymetrix GeneChip Mouse Genome 430 2.0 Array |
71182_PM_at |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR0300003056.mm.1 |
NR_033592.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300003056.mm.1 |
NR_033592.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300003056.mm.1 |
TC0300000434.mm.1 |
MTA TC |
Mouse Expression Set 430 |
1454539_PM_at |
- |
- |
Mouse Expression Set 430 |
1433280_PM_at |
- |
- |
Mouse Expression Set 430 |
1454539_at |
- |
- |
Mouse Expression Set 430 |
1433280_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1454539_PM_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1433280_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1454539_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1433280_PM_at |
- |
- |