Assay Details
Gene Symbol: Sohlh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028937.3 | NP_083213.2 | 1 - 2 | 68 | 81 | |
GenBank mRNA | BC120917.1 | - | 1 - 2 | 63 | 40092728 | 81 |
BC120918.2 | - | 1 - 2 | 63 | 40092729 | 81 | |
DQ086115.1 | - | 1 - 2 | 68 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
