Assay Details
Gene Symbol: Fabp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008375.2 | NP_032401.1 | 1 - 2 | 115 | 123 | |
GenBank mRNA | BC119289.1 | - | 1 - 2 | 85 | 8734038 | 123 |
BC120767.1 | - | 1 - 2 | 85 | 40129690 | 123 | |
CJ043022.1 | - | 1 - 2 | 115 | 123 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
