Assay Details
| Gene Symbol: Brms1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037756.2 | NP_001032845.1 | 2 - 3 | 361 | 101 | |
| GenBank mRNA | AK003055.1 | - | 2 - 3 | 110 | 101 | |
| AK154678.1 | - | 2 - 3 | 362 | 101 | ||
| AK155742.1 | - | 2 - 3 | 362 | 101 | ||
| BC034885.1 | - | 2 - 3 | 322 | 1362264 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Brms1l
Genomic Map