Assay Details
| Gene Symbol: Brms1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037756.2 | NP_001032845.1 | 7 - 8 | 825 | 70 | |
| GenBank mRNA | AK154678.1 | - | 7 - 8 | 826 | 70 | |
| AK155742.1 | - | 7 - 8 | 826 | 70 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Brms1l
Genomic Map