Assay Details
Gene Symbol: Trove2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013835.2 | NP_038863.1 | 4 - 5 | 1101 | 122 | |
GenBank mRNA | AK076167.1 | - | 4 - 5 | 1078 | 122 | |
AK167379.1 | - | 4 - 5 | 1042 | 122 | ||
AK167555.1 | - | 4 - 5 | 1042 | 122 | ||
AK167645.1 | - | 4 - 5 | 1042 | 122 | ||
BC040368.1 | - | 3 - 4 | 972 | 122 | ||
BC051974.1 | - | 3 - 4 | 978 | 5697399 | 122 | |
BC052380.1 | - | 3 - 4 | 978 | 5697399 | 122 | |
L81154.1 | - | 3 - 4 | 949 | 122 | ||
U66843.1 | - | 4 - 5 | 1028 | 122 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
