Assay Details
| Gene Symbol: Rgs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009061.4 | NP_033087.2 | 3 - 4 | 311 | 111 | |
| GenBank mRNA | AF215668.1 | - | 3 - 4 | 296 | 111 | |
| AF432916.1 | - | 3 - 4 | 290 | 111 | ||
| AK029310.1 | - | 3 - 4 | 313 | 111 | ||
| AK031603.1 | - | 3 - 4 | 313 | 111 | ||
| AK077922.1 | - | 3 - 4 | 312 | 111 | ||
| AK162276.1 | - | 3 - 4 | 313 | 111 | ||
| BC023001.1 | - | 3 - 4 | 294 | 5365271 | 111 | |
| U67187.1 | - | 3 - 4 | 300 | 111 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rgs2
Genomic Map