Assay Details
| Gene Symbol: Sec23a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009147.2 | NP_033173.2 | 14 - 15 | 1851 | 84 | |
| GenBank mRNA | AK028152.1 | - | 14 - 15 | 1739 | 84 | |
| AY082671.1 | - | 14 - 15 | 1725 | 84 | ||
| BC034610.1 | - | 14 - 15 | 1851 | 5364796 | 84 | |
More Information
Set Membership: |
|

Sec23a
Genomic Map