Assay Details
Gene Symbol: Sec23a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009147.2 | NP_033173.2 | 16 - 17 | 2100 | 78 | |
GenBank mRNA | AK028152.1 | - | 16 - 17 | 1988 | 78 | |
AY082671.1 | - | 16 - 17 | 1974 | 78 | ||
BC034610.1 | - | 16 - 17 | 2100 | 5364796 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
