Assay Details
| Gene Symbol: Maob | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172778.2 | NP_766366.2 | 5 - 6 | 567 | 80 | |
| GenBank mRNA | AK031833.1 | - | 5 - 6 | 551 | 80 | |
| AK054050.1 | - | 5 - 6 | 566 | 80 | ||
| BC113182.1 | - | 5 - 6 | 556 | 40061991 | 80 | |
| BC113788.1 | - | 5 - 6 | 556 | 40061990 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Maob
Genomic Map