Assay Details
| Gene Symbol: Maoa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173740.3 | NP_776101.3 | 2 - 3 | 308 | 71 | |
| GenBank mRNA | AK004337.1 | - | 2 - 3 | 170 | 71 | |
| AK146612.1 | - | 2 - 3 | 309 | 71 | ||
| AK155052.1 | - | 2 - 3 | 171 | 71 | ||
| AK164457.1 | - | 2 - 3 | 145 | 71 | ||
| BC029100.1 | - | 2 - 3 | 178 | 3481702 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Maoa
Genomic Map