Assay Details
| Gene Symbol: Tcf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009331.3 | NP_033357.1 | 8 - 9 | 829 | 102 | |
| GenBank mRNA | AK037616.1 | - | 8 - 9 | 830 | 102 | |
| BC056630.1 | - | 7 - 8 | 1071 | 102 | ||
| BC138596.1 | - | 7 - 8 | 809 | 8861616 | 102 | |
| BC145343.1 | - | 7 - 8 | 809 | 9053899 | 102 | |
| X61385.1 | - | 9 - 10 | 971 | 102 | ||
More Information
Set Membership: |
|

Tcf7
Genomic Map