Assay Details
Gene Symbol: Lrrc34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027941.1 | NP_082217.1 | 5 - 6 | 534 | 102 | |
GenBank mRNA | AK005720.1 | - | 5 - 6 | 586 | 102 | |
BC100519.1 | - | 5 - 6 | 534 | 30919449 | 102 | |
BC116419.1 | - | 4 - 5 | 367 | 102 | ||
BC116420.1 | - | 4 - 5 | 374 | 40103399 | 102 | |
JF934738.1 | - | 5 - 6 | 485 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
