Assay Details
| Gene Symbol: Prkci | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008857.3 | NP_032883.2 | 16 - 17 | 1796 | 152 | |
| GenBank mRNA | AK144979.1 | - | 16 - 17 | 1796 | 152 | |
| AK167411.1 | - | 16 - 17 | 1765 | 152 | ||
| AK220517.1 | - | 16 - 17 | 1820 | 152 | ||
| BC021630.1 | - | 16 - 17 | 1667 | 3980628 | 152 | |
| D28577.1 | - | 16 - 17 | 1591 | 152 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prkci
Genomic Map