Assay Details
| Gene Symbol: Syt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009307.3 | NP_033333.2 | 5 - 6 | 849 | 89 | |
| GenBank mRNA | AF257304.1 | - | 4 - 5 | 604 | 89 | |
| AK036357.1 | - | 5 - 6 | 850 | 89 | ||
| AK220537.1 | - | 2 - 3 | 163 | 89 | ||
| BC027019.1 | - | 4 - 5 | 610 | 5362352 | 89 | |
| D37793.1 | - | 3 - 4 | 483 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Syt2
Genomic Map