Assay Details
| Gene Symbol: Cdhr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024478.1 | NP_001019649.1 | 8 - 9 | 1152 | 74 | |
| GenBank mRNA | AK047666.1 | - | 8 - 9 | 1153 | 74 | |
| AK049806.1 | - | 8 - 9 | 1154 | 74 | ||
| BC138911.1 | - | 8 - 9 | 1080 | 8861933 | 74 | |
More Information
Set Membership: |
|

Cdhr3
Genomic Map