Assay Details
| Gene Symbol: Zfp639 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161818.1 | NP_001155290.1 | 1 - 2 | 45 | 97 | |
| GenBank mRNA | AK078000.1 | - | 1 - 2 | 46 | 97 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK078000.1, NM_001161818.1 transcripts.
Set Membership: |
|

Zfp639
Genomic Map