Assay Details
| Gene Symbol: Akap5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101471.1 | NP_001094941.1 | 2 | 2384 | 119 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of protein phosphorylation
protein targeting
signal transduction
negative regulation of adenylate cyclase activity
positive regulation of calcium ion transport into cytosol
positive regulation of protein kinase A signaling
positive regulation of cAMP biosynthetic process
amylase secretion
negative regulation of potassium ion transport
negative regulation of ion transport
cellular protein complex disassembly
positive regulation of adenylate cyclase activity
positive regulation of dendrite morphogenesis
positive regulation of NFAT protein import into nucleus
G-protein coupled receptor binding
actin binding
protein binding
calmodulin binding
adenylate cyclase binding
kinase binding
protein kinase binding
protein domain specific binding
protein phosphatase 2B binding
protein complex binding
protein kinase A regulatory subunit binding
cadherin binding
GABA receptor binding
positive regulation of protein phosphorylation
protein targeting
signal transduction
negative regulation of adenylate cyclase activity
positive regulation of calcium ion transport into cytosol
positive regulation of protein kinase A signaling
positive regulation of cAMP biosynthetic process
amylase secretion
negative regulation of potassium ion transport
negative regulation of ion transport
cellular protein complex disassembly
positive regulation of adenylate cyclase activity
positive regulation of dendrite morphogenesis
positive regulation of NFAT protein import into nucleus
G-protein coupled receptor binding
actin binding
protein binding
calmodulin binding
adenylate cyclase binding
kinase binding
protein kinase binding
protein domain specific binding
protein phosphatase 2B binding
protein complex binding
protein kinase A regulatory subunit binding
cadherin binding
GABA receptor binding
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P1042732 | NM_001101471.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P1042732 | AK014386.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P1042732 | NM_001101471.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P1042732 | AK014386.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 566753 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 272441 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 758772 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 894740 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 341093 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 781144 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 126247 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 70397 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P1042732 | NM_001101471.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P1042732 | AK014386.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P1042732 | NM_001101471.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P1042732 | AK014386.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P1042732 | NM_001101471.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P1042732 | AK014386.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P1042732 | NM_001101471.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P1042732 | AK014386.1 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28553329 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10160045 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553375 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553373 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553367 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553336 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10317306 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553276 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553320 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553287 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553300 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553316 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553341 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553295 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553339 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553313 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553278 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10410875 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553310 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553365 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10115814 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553357 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553361 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10272552 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10317298 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553303 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553382 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553362 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553331 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553348 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10115813 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553378 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10326591 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553289 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553329 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10317292 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553308 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553377 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553277 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553383 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553351 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553359 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553328 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553345 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10115804 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10317300 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553296 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10272558 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10018521 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553302 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553350 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553337 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553311 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553374 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553379 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553326 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553317 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553279 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553376 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553354 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553292 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553364 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553347 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553353 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28553282 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10396643 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10396641 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10396642 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10396644 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1447301_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1431717_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1436142_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10396640 | - | - |
| Mouse Genome 430 2.0 Array | 1436142_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1431717_at | - | - |
| Mouse Genome 430 2.0 Array | 1431717_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1436142_at | - | - |
| Mouse Genome 430 2.0 Array | 1447301_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1447301_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 238276_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 238276_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1200004804.mm.1 | NM_001101471.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200004804.mm.1 | NM_001101471.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200004804.mm.1 | TC1200000676.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200004801.mm.1 | NM_001101471.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200004801.mm.1 | NM_001101471.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200004801.mm.1 | TC1200000676.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200004805.mm.1 | NM_001101471.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200004805.mm.1 | NM_001101471.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200004805.mm.1 | TC1200000676.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200004803.mm.1 | NM_001101471.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200004803.mm.1 | NM_001101471.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200004803.mm.1 | TC1200000676.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1431717_PM_at | - | - |
| Mouse Expression Set 430 | 1447301_PM_at | - | - |
| Mouse Expression Set 430 | 1447301_at | - | - |
| Mouse Expression Set 430 | 1436142_at | - | - |
| Mouse Expression Set 430 | 1436142_PM_at | - | - |
| Mouse Expression Set 430 | 1431717_at | - | - |
| Murine Genome U74v2 Set | 137165_PM_at | - | - |
| Murine Genome U74v2 Set | 137165_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431717_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431717_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1447301_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1436142_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1436142_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1447301_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 866474 | - | - |
| ABI Mouse Genome Survey Microarray | 877625 | - | - |

Akap5
Genomic Map