Assay Details
| Gene Symbol: Dsel | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081316.1 | NP_001074785.1 | 1 - 2 | 820 | 150 | |
| GenBank mRNA | AK033983.1 | - | 1 - 2 | 821 | 150 | |
| AK164854.1 | - | 1 - 2 | 670 | 150 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK033983.1, NM_001081316.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20528693 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20528694 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20528697 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20528696 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20528695 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P660693 | AK164854.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P660693 | BC120567.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P660693 | BC137796.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P660693 | AK032110.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P660693 | NM_001081316.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P660693 | AK164854.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P660693 | BC120567.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P660693 | BC137796.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P660693 | AK032110.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P660693 | NM_001081316.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 507115 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 671924 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 945265 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 71615 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 30927 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P112999 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P139515 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P660693 | AK164854.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P660693 | BC120567.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P660693 | BC137796.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P660693 | AK032110.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P660693 | NM_001081316.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P660693 | AK164854.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P660693 | BC120567.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P660693 | BC137796.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P660693 | AK032110.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P660693 | NM_001081316.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P139515 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P112999 | - | - |
| Whole Mouse Genome G4122A | A_51_P183723 | AK033983.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P183723 | BC120567.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P183723 | BC137796.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P183723 | AK032110.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P183723 | NM_001081316.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P39056 | AK033983.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P39056 | AK164854.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P39056 | NM_001081316.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P660693 | AK164854.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P660693 | BC120567.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P660693 | BC137796.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P660693 | AK032110.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P660693 | NM_001081316.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P660693 | AK164854.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P660693 | BC120567.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P660693 | BC137796.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P660693 | AK032110.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P660693 | NM_001081316.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P112999 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P139515 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20528694 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20528693 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20528695 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20528697 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20528696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20528757 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528731 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528725 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528732 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528643 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528753 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01008671 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528662 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528710 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528635 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528645 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528632 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01008689 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528683 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528671 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528735 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528649 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528636 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00588262 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528652 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528680 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528661 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528679 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528641 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528634 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528660 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528742 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528666 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528726 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528752 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528670 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528751 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528637 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528745 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528738 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528640 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528749 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528758 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528657 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528676 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528703 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528711 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528713 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528706 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528722 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528668 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528750 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528716 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528648 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528659 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528655 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528727 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00588239 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01008702 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528708 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528677 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00315937 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528695 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528689 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528704 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00151076 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528718 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528754 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528715 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528729 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528712 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528705 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528685 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528681 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528748 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00279291 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528709 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528739 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528647 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20528691 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10357118 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10357117 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10357116 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10357119 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1438407_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10357115 | - | - |
| Mouse Genome 430 2.0 Array | 1438407_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1438407_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 319901_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 319901_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100023562.mm.1 | NM_001081316.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100023562.mm.1 | NM_001081316.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100023562.mm.1 | TC0100002921.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100023563.mm.1 | NM_001081316.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100023563.mm.1 | NM_001081316.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100023563.mm.1 | TC0100002921.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100023564.mm.1 | NM_001081316.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100023564.mm.1 | NM_001081316.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100023564.mm.1 | TC0100002921.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100023565.mm.1 | NM_001081316.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100023565.mm.1 | NM_001081316.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100023565.mm.1 | TC0100002921.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1438407_at | - | - |
| Mouse Expression Set 430 | 1438407_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1438407_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1438407_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 642518 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1222968 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1222968 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2956387 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2956387 | - | - |

Dsel
Genomic Map