Assay Details
| Gene Symbol: Sar1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025535.2 | NP_079811.1 | 1 - 2 | 130 | 88 | |
| GenBank mRNA | AK002327.1 | - | 1 - 2 | 107 | 88 | |
| AK150906.1 | - | 1 - 2 | 86 | 88 | ||
| AK155429.1 | - | 1 - 2 | 100 | 88 | ||
| BC082550.1 | - | 1 - 2 | 99 | 30357800 | 88 | |
| BC085178.1 | - | 1 - 2 | 83 | 30701201 | 88 | |
| BY068863.1 | - | 1 - 2 | 130 | 88 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK002327.1, AK150906.1, AK155429.1, BC082550.1, BC085178.1, NM_025535.2 transcripts.
Set Membership: |
|

Sar1b
Genomic Map