Assay Details
| Gene Symbol: Sar1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025535.2 | NP_079811.1 | 5 - 6 | 497 | 76 | |
| GenBank mRNA | AK002327.1 | - | 5 - 6 | 474 | 76 | |
| AK013559.1 | - | 5 - 6 | 459 | 76 | ||
| AK150906.1 | - | 5 - 6 | 453 | 76 | ||
| AK155429.1 | - | 5 - 6 | 467 | 76 | ||
| BC082550.1 | - | 5 - 6 | 466 | 30357800 | 76 | |
| BC085178.1 | - | 5 - 6 | 450 | 30701201 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sar1b
Genomic Map