Assay Details
| Gene Symbol: Sec24a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290785.1 | NP_001277714.1 | 8 - 9 | 2028 | 143 | |
| NM_175255.3 | NP_780464.2 | 8 - 9 | 2031 | 143 | ||
| GenBank mRNA | AK029139.1 | - | 8 - 9 | 1784 | 143 | |
| AK155182.1 | - | 8 - 9 | 1817 | 143 | ||
| AK162795.1 | - | 8 - 9 | 1636 | 143 | ||
| AK163998.1 | - | 8 - 9 | 1625 | 143 | ||
| BC117905.1 | - | 8 - 9 | 1295 | 143 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sec24a
Genomic Map