Assay Details
| Gene Symbol: Snrpe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009227.3 | NP_033253.1 | 4 - 5 | 281 | 130 | |
| GenBank mRNA | AK019462.1 | - | 4 - 5 | 265 | 130 | |
| AW492281.1 | - | 1 - 2 | 130 | |||
| BC008262.1 | - | 4 - 5 | 257 | 2650413 | 130 | |
| BC051207.1 | - | 4 - 5 | 239 | 1396204 | 130 | |
| BC055765.1 | - | 4 - 5 | 223 | 6407944 | 130 | |
| BY168551.1 | - | 4 - 5 | 281 | 130 | ||
| X65704.1 | - | 4 - 5 | 185 | 130 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Snrpe
Genomic Map