Assay Details
Gene Symbol: Gmps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033300.2 | NP_001028472.2 | 3 - 4 | 465 | 102 | |
GenBank mRNA | AK049755.1 | - | 3 - 4 | 429 | 102 | |
AK143197.1 | - | 3 - 4 | 466 | 102 | ||
AK146654.1 | - | 3 - 4 | 483 | 102 | ||
AK167857.1 | - | 3 - 4 | 445 | 102 | ||
AK168239.1 | - | 3 - 4 | 415 | 102 | ||
AK169043.1 | - | 3 - 4 | 449 | 102 | ||
AK169701.1 | - | 3 - 4 | 463 | 102 | ||
BC037232.1 | - | 1 - 2 | 117 | 102 | ||
BC138411.1 | - | 3 - 4 | 421 | 8861431 | 102 | |
BC138412.1 | - | 3 - 4 | 421 | 8861432 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
