Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200002726.mm.1 |
NM_026164.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200002726.mm.1 |
NM_026164.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200002726.mm.1 |
TC1200000387.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200002722.mm.1 |
NM_026164.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200002722.mm.1 |
NM_026164.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200002722.mm.1 |
TC1200000387.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200002721.mm.1 |
NM_026164.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200002721.mm.1 |
NM_026164.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200002721.mm.1 |
TC1200000387.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200002727.mm.1 |
NM_026164.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200002727.mm.1 |
NM_026164.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200002727.mm.1 |
TC1200000387.mm.1 |
MTA TC |