Assay Details
| Gene Symbol: Pnpla8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026164.2 | NP_080440.2 | 5 - 6 | 1668 | 97 | |
| GenBank mRNA | AB044139.1 | - | 5 - 6 | 1547 | 97 | |
| AK004621.1 | - | 5 - 6 | 1635 | 97 | ||
| AK084928.1 | - | 1335 | 97 | |||
| AK145776.1 | - | 5 - 6 | 1578 | 97 | ||
| AK163211.1 | - | 5 - 6 | 1636 | 97 | ||
| AK168475.1 | - | 5 - 6 | 1631 | 97 | ||
| BC019364.1 | - | 2 - 3 | 122 | 97 | ||
| BC127056.1 | - | 4 - 5 | 1528 | 40109753 | 97 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC019364.1 transcripts.
Set Membership: |
|

Pnpla8
Genomic Map