Assay Details
| Gene Symbol: Hnf4g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013920.2 | NP_038948.1 | 5 - 6 | 840 | 75 | |
| GenBank mRNA | AJ242626.1 | - | 5 - 6 | 792 | 75 | |
| AK143816.1 | - | 5 - 6 | 841 | 75 | ||
| BC125519.1 | - | 5 - 6 | 662 | 40130034 | 75 | |
More Information
Set Membership: |
|

Hnf4g
Genomic Map