Assay Details
| Gene Symbol: Hnf4g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013920.2 | NP_038948.1 | 6 - 7 | 924 | 79 | |
| GenBank mRNA | AJ242626.1 | - | 6 - 7 | 876 | 79 | |
| AK143816.1 | - | 6 - 7 | 925 | 79 | ||
| BC125519.1 | - | 6 - 7 | 746 | 40130034 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hnf4g
Genomic Map