Assay Details
Gene Symbol: Hnf4g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013920.2 | NP_038948.1 | 8 - 9 | 1316 | 69 | |
GenBank mRNA | AJ242626.1 | - | 8 - 9 | 1268 | 69 | |
AK143816.1 | - | 8 - 9 | 1317 | 69 | ||
BC125519.1 | - | 8 - 9 | 1138 | 40130034 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
