Assay Details
| Gene Symbol: Pxdn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181395.2 | NP_852060.2 | 17 - 18 | 3693 | 119 | |
| GenBank mRNA | AK122223.1 | - | 17 - 18 | 3463 | 119 | |
| AK142872.1 | - | 17 - 18 | 3693 | 119 | ||
| BC037709.1 | - | 1 - 2 | 610 | 119 | ||
| BC044828.1 | - | 2 - 3 | 1657 | 119 | ||
| BC049858.1 | - | 2 - 3 | 1657 | 119 | ||
| BC112913.1 | - | 8 - 9 | 2490 | 119 | ||
| BC150789.1 | - | 17 - 18 | 3688 | 9087700 | 119 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pxdn
Genomic Map