Assay Details
| Gene Symbol: Tpo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009417.2 | NP_033443.1 | 5 - 6 | 537 | 82 | |
| GenBank mRNA | AK047843.1 | - | 5 - 6 | 516 | 82 | |
| X60703.1 | - | 5 - 6 | 537 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tpo
Genomic Map