Assay Details
| Gene Symbol: Swt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025819.4 | NP_080095.3 | 2 - 3 | 232 | 92 | |
| GenBank mRNA | AK004803.2 | - | 2 - 3 | 179 | 92 | |
| BC028449.1 | - | 2 - 3 | 160 | 92 | ||
| BC055064.1 | - | 2 - 3 | 233 | 6528686 | 92 | |
| BC060204.1 | - | 2 - 3 | 172 | 6855442 | 92 | |
| BY062718.1 | - | 2 - 3 | 232 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Swt1
Genomic Map