Assay Details
| Gene Symbol: 2610027F03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK011565.1 | - | 6 - 7 | 842 | 60 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | engrailed 1 | En1 |
En-1, Mo-en.1 |
-81.1kb | Sense | Chr. 1 - 120602391 - 120607991 on NCBI Build GRCm38 |
| Downstream | cerebellum expressed regulatory RNA | Celrr |
AI848258, B230209K01Rik, Celr, Gm818 |
397.0kb | Sense | Chr. 1 - 121087405 - 121120975 on NCBI Build GRCm38 |
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2274961 | AK011565.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 15173 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 328222 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 807600 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 643651 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 843859 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 372072 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 645559 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 588084 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 626341 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608737 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924764 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 921553 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 257643 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 932475 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 109404 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 837536 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 236849 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 731717 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 771463 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 264026 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 181131 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 891142 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 240894 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 830902 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 333443 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 838805 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 101156 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 165568 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 98830 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 40946 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 102003 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 75843 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P112245 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P113197 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P126388 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P126388 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P113197 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P112245 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04240549 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04240531 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04240584 | - | - |
| Whole Mouse Genome G4122A | A_52_P243992 | AK011565.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P126388 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P112245 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P04240584 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P113197 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00635309 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00703196 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00263245 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20562917 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00256562 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20562948 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20562890 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00779403 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20563007 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20563055 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20563035 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20562906 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00030647 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20563062 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20562893 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00030648 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20562932 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20563086 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20563039 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00784237 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20563082 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00174713 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00765218 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01067283 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01067285 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20563030 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00580509 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20563049 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00703185 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20563044 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20562910 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00790227 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20562888 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20562923 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00256569 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00737469 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20563072 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20563050 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20563041 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00271268 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20563056 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20563004 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00703183 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00874457 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20562942 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20562891 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20562899 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20562930 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00030799 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20562912 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20562937 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20563089 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20563002 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00784236 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20563006 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00263250 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20563059 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20562905 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00262751 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20563079 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00784230 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20562884 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20563048 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20562898 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20563012 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01017805 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20562925 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20563070 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00632160 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20562920 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00256574 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20563066 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20562887 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20562914 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20563060 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20562895 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20562911 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20562881 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00784224 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00523895 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00384792 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20563038 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20563063 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00635308 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT MG-430 PM Array Plate | 1454178_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1454178_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1454178_at | - | - |
| Mouse Expression Set 430 | 1454178_PM_at | - | - |
| Mouse Expression Set 430 | 1454178_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1454178_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1454178_at | - | - |

2610027F03Rik
Genomic Map