Assay Details
Gene Symbol: Ids | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010498.3 | NP_034628.2 | 4 - 5 | 702 | 81 | |
GenBank mRNA | AJ627044.1 | - | 2 - 3 | 167 | 81 | |
AK085237.1 | - | 5 - 6 | 579 | 81 | ||
AK132287.1 | - | 5 - 6 | 582 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
