Assay Details
| Gene Symbol: Zc2hc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173181.3 | NP_775273.1 | 1 - 2 | 102 | 67 | |
| GenBank mRNA | AK014202.1 | - | 1 - 2 | 38 | 67 | |
| AK083941.1 | - | 1 - 2 | 46 | 67 | ||
| AK085187.1 | - | 1 - 2 | 39 | 67 | ||
| AK170609.1 | - | 1 - 2 | 43 | 67 | ||
| BC087731.1 | - | 66 | 6850296 | 67 | ||
| BY131077.1 | - | 1 - 2 | 103 | 67 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK083941.1, AK170609.1, BC087731.1, NM_173181.3 transcripts.
Set Membership: |
|

Zc2hc1a
Genomic Map