Assay Details
| Gene Symbol: Zc2hc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173181.3 | NP_775273.1 | 5 - 6 | 587 | 130 | |
| GenBank mRNA | AK014202.1 | - | 5 - 6 | 523 | 130 | |
| AK083941.1 | - | 5 - 6 | 531 | 130 | ||
| AK085187.1 | - | 5 - 6 | 525 | 130 | ||
| AK170609.1 | - | 5 - 6 | 528 | 130 | ||
| BC044809.1 | - | 5 - 6 | 509 | 130 | ||
| BC087731.1 | - | 551 | 6850296 | 130 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zc2hc1a
Genomic Map