Assay Details
| Gene Symbol: Cenpl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK053363.1 | - | 5 - 6 | 1413 | 91 | |
| AK150539.1 | - | 5 - 6 | 1572 | 91 | ||
| AK151163.1 | - | 5 - 6 | 1572 | 91 | ||
| BC048708.1 | - | 4 - 5 | 1203 | 91 | ||
| BC063765.1 | - | 4 - 5 | 1246 | 91 | ||
| BC080823.1 | - | 4 - 5 | 1256 | 91 | ||
| BC116944.1 | - | 4 - 5 | 1354 | 40126263 | 91 | |
| BC116970.1 | - | 4 - 5 | 1354 | 40126289 | 91 | |
| BC144746.1 | - | 4 - 5 | 1255 | 91 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20769881 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20769882 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20769883 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P17827 | AK154611.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P17827 | BC060191.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P17827 | AK028960.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P17827 | NM_027429.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2077228 | BC144746.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2077228 | BC063765.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2077228 | BC080823.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2077228 | BC116944.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2077228 | BC116970.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 909544 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 144955 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 293335 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 297836 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 459028 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 56812 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 456152 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 258068 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 284081 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 127400 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P155253 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P147431 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P17827 | AK154611.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P17827 | BC060191.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P17827 | AK028960.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P17827 | NM_027429.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2077228 | BC144746.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2077228 | BC063765.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2077228 | BC080823.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2077228 | BC116944.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2077228 | BC116970.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20769898 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20769887 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20769900 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20769894 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20769886 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20769904 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20769906 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20769895 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20769888 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20769892 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20769890 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20769908 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20769893 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20769910 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20769881 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20769905 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20769914 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20769884 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20769902 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04329913 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P147431 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P155253 | - | - |
| Whole Mouse Genome G4122A | A_51_P195808 | BC020064.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P362997 | BC144746.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P362997 | BC063765.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P362997 | BC080823.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P362997 | BC116944.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P362997 | BC116970.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P147431 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P155253 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00202776 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00202787 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00202789 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20769883 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20769881 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20769882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20769898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20769956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20769889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20769892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20769884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20769902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20769943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20769887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20769906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20769904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20769939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20769880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20769923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20769938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20769935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20769947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20769895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20769890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20769945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20769927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20769886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20769894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20769957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20769958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20769953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20769881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20769942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20769930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20769888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20769900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20769949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20769893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20769932 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20769910 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20769895 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01014873 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20769881 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20769914 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20769898 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20769890 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01061627 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20769908 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00508759 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20769919 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20769905 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00740644 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20769906 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20769880 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20769886 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20769911 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20769887 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01092382 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20769923 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20769917 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20769902 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00740651 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20769921 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20769893 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20769888 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20769915 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00740647 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20769918 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20769884 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20769900 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10351049 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351053 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351055 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351051 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351048 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351050 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351054 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10351052 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429326_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10351047 | - | - |
| Mouse Genome 430 2.0 Array | 1429326_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1429326_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70454_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70454_PM_at | - | - |
| Mouse Expression Set 430 | 1429326_at | - | - |
| Mouse Expression Set 430 | 1429326_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429326_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429326_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1254468 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2676726 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2883414 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2883414 | - | - |

Cenpl
Genomic Map