Assay Details
Gene Symbol: Gpr101 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033360.3 | NP_001028532.1 | 1 - 2 | 221 | 58 | |
GenBank mRNA | AK138185.1 | - | 1 - 2 | 165 | 58 | |
AK138370.1 | - | 1 - 2 | 223 | 58 | ||
DQ198079.1 | - | 1 - 2 | 181 | 58 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK138185.1, AK138370.1, DQ198079.1, NM_001033360.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32401473 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32401470 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32401469 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32401472 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32401468 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2011600 | AK033976.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2011600 | DQ198079.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2011600 | NM_001033360.3 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 167559 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 393025 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 147834 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 654083 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 115721 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P153219 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2011600 | AK033976.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2011600 | DQ198079.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2011600 | NM_001033360.3 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32401467 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32401482 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32401484 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32401499 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32401490 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32401501 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32401476 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32401494 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32401479 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32401459 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32401468 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32401496 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32401464 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32401472 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32401489 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32401493 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32401462 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32401473 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32401477 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32401452 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32401465 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32401454 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32401470 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32401491 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32401478 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32401483 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32401495 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32401458 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32401460 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32401463 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32401487 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32401456 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32401497 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32401492 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32401488 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32401481 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32401474 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P153219 | - | - |
Whole Mouse Genome G4122A | A_52_P299958 | AK138185.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P299958 | AK138370.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P299958 | AY255576.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P299958 | BC139013.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P299958 | BC139014.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P153219 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32401472 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32401468 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32401470 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32401473 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32401469 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401478 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401464 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401477 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401443 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401511 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401492 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401523 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401473 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401426 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401456 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401458 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401412 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401435 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401510 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401459 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401494 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401472 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401413 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401462 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401408 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401476 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401446 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401444 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401488 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401504 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401513 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401430 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401470 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401483 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401399 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401450 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401490 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401405 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401389 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401518 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401393 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401479 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401398 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401401 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401440 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401424 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401487 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401403 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401460 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401520 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401417 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401465 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401506 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401463 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401514 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401451 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401395 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401421 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401497 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401495 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401382 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401423 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401499 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401410 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401404 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401386 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401468 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401432 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401502 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401407 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401449 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401522 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401527 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401474 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401437 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401447 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401496 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401454 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401491 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401517 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401419 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401390 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401525 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401507 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401415 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401526 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32401416 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401483 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401524 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P15017391 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401496 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401428 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P15041807 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401482 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401473 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P15041803 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401512 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P15146914 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P15041802 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401426 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401450 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401488 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401432 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401502 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401506 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401516 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14948556 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401520 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401437 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401467 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P15041795 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401526 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401478 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P15026266 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14825448 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401463 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401489 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401479 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P15017389 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401525 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14871560 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401494 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401507 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401518 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401415 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401474 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401477 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401424 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P15041801 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401446 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14871567 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401499 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401410 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401527 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401458 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401443 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14712271 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401440 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401513 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401417 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401504 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P15041794 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401514 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401454 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401430 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401419 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401435 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401421 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401409 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14712274 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401412 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401476 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401490 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P15041800 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401497 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401413 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401522 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401501 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401456 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P15041789 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401510 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401433 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401423 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32401416 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10604747 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10604746 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10604748 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1441382_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10604745 | - | - |
Mouse Genome 430 2.0 Array | 1441382_at | - | - |
Mouse Genome 430 2.0 Array | 1441382_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 245424_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 245424_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0X00014456.mm.1 | NM_001033360.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00014456.mm.1 | NM_001033360.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00014456.mm.1 | TC0X00002274.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0X00014458.mm.1 | NM_001033360.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00014458.mm.1 | NM_001033360.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00014458.mm.1 | TC0X00002274.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0X00014455.mm.1 | NM_001033360.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00014455.mm.1 | NM_001033360.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00014455.mm.1 | TC0X00002274.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0X00014453.mm.1 | NM_001033360.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00014453.mm.1 | NM_001033360.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00014453.mm.1 | TC0X00002274.mm.1 | MTA TC |
Mouse Expression Set 430 | 1441382_PM_at | - | - |
Mouse Expression Set 430 | 1441382_at | - | - |
Murine Genome U74v2 Set | 135331_at | - | - |
Murine Genome U74v2 Set | 135331_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1441382_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1441382_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 333022 | - | - |
ABI Mouse Genome Survey Microarray | 364871 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.1 expression beadchip | ILMN_1258301 | - | - |
