Assay Details
Gene Symbol: Gm9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033234.2 | NP_001028406.1 | 3 - 4 | 629 | 69 | |
GenBank mRNA | AK131663.1 | - | 3 - 4 | 594 | 69 | |
AK145837.1 | - | 3 - 4 | 630 | 69 | ||
BC048624.1 | - | 3 - 4 | 593 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P127206 | BC048624.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P127206 | NM_001033234.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P127206 | AK131663.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P127206 | AK145837.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P127206 | BC048624.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P127206 | NM_001033234.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P127206 | AK131663.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P127206 | AK145837.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 302375 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P127206 | BC048624.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P127206 | NM_001033234.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P127206 | AK131663.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P127206 | AK145837.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P127206 | BC048624.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P127206 | NM_001033234.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P127206 | AK131663.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P127206 | AK145837.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09265746 | - | - |
Whole Mouse Genome G4122A | A_52_P127206 | BC048624.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P127206 | NM_001033234.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P127206 | AK131663.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P127206 | AK145837.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P127206 | BC048624.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P127206 | NM_001033234.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P127206 | AK131663.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P127206 | AK145837.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09265746 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322167 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322177 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322242 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322198 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322206 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322213 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322153 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322175 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322160 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322164 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322220 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322262 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322244 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322246 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322201 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322202 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322240 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322217 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322222 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322192 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322260 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322191 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322189 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322173 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322215 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322179 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322159 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322218 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322258 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322208 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322247 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322212 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322190 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322204 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322158 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322214 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322251 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322186 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322168 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322239 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322194 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322254 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322188 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322241 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322163 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322157 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32322176 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32322196 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32322173 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32322192 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14927434 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32322195 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32322197 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32322189 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32322202 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32322206 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P15075479 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32322217 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P15075478 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14927432 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32322222 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32322218 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32322194 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P15075485 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32322177 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32322179 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32322203 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14939975 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32322190 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32322213 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32322215 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32322186 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32322201 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32322171 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P15075484 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14556528 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14882948 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32322184 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32322168 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32322208 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P15075474 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32322214 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14759716 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32322182 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32322167 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32322175 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10604107 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10604109 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10604108 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10604110 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1443995_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10604106 | - | - |
Mouse Genome 430 2.0 Array | 1443995_at | - | - |
Mouse Genome 430 2.0 Array | 1443995_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 194854_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 194854_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0X00013119.mm.1 | NM_001033234.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00013119.mm.1 | NM_001033234.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00013119.mm.1 | TC0X00002062.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0X00013120.mm.1 | NM_001033234.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00013120.mm.1 | NM_001033234.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00013120.mm.1 | TC0X00002062.mm.1 | MTA TC |
Mouse Expression Set 430 | 1443995_at | - | - |
Mouse Expression Set 430 | 1443995_PM_at | - | - |
Murine Genome U74v2 Set | 170846_PM_at | - | - |
Murine Genome U74v2 Set | 170846_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1443995_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1443995_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 888987 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.1 expression beadchip | ILMN_1217112 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2931123 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2931123 | - | - |
