Assay Details
| Gene Symbol: Pcbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103165.1 | NP_001096635.1 | 1 - 2 | 172 | 80 | |
| NM_001103166.1 | NP_001096636.1 | 1 - 2 | 172 | 80 | ||
| NM_011042.2 | NP_035172.2 | 1 - 2 | 172 | 80 | ||
| GenBank mRNA | AK169268.1 | - | 1 - 2 | 266 | 80 | |
| L19661.1 | - | 1 - 2 | 59 | 80 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK169268.1,L19661.1,NM_001103165.1,NM_001103166.1,NM_011042.2 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Pcbp2
Genomic Map