Assay Details
| Gene Symbol: Eif4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145625.3 | NP_663600.2 | 12 - 13 | 1756 | 87 | |
| GenBank mRNA | AK045250.1 | - | 12 - 13 | 1600 | 87 | |
| AK077423.1 | - | 12 - 13 | 1607 | 87 | ||
| AK148012.1 | - | 12 - 13 | 1606 | 87 | ||
| AK161693.1 | - | 12 - 13 | 1589 | 87 | ||
| AK163266.1 | - | 12 - 13 | 1607 | 87 | ||
| AK166363.1 | - | 12 - 13 | 1608 | 87 | ||
| AK168350.1 | - | 12 - 13 | 1602 | 87 | ||
| AK168425.1 | - | 12 - 13 | 1602 | 87 | ||
| AK170256.1 | - | 12 - 13 | 1607 | 87 | ||
| AK170354.1 | - | 12 - 13 | 1589 | 87 | ||
| AK178737.1 | - | 3 - 4 | 378 | 87 | ||
| AK179593.1 | - | 3 - 4 | 346 | 87 | ||
| AK211186.1 | - | 3 - 4 | 320 | 87 | ||
| AK219884.1 | - | 3 - 4 | 334 | 87 | ||
| BC007171.1 | - | 11 - 12 | 1555 | 3492114 | 87 | |
| BC036293.1 | - | 12 - 13 | 1592 | 87 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Eif4b
Genomic Map