Assay Details
| Gene Symbol: Aaas | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153416.2 | NP_700465.2 | 7 - 8 | 831 | 69 | |
| GenBank mRNA | AK034591.1 | - | 7 - 8 | 726 | 69 | |
| AK079849.1 | - | 7 - 8 | 768 | 69 | ||
| AK083401.1 | - | 7 - 8 | 744 | 69 | ||
| AK083537.1 | - | 7 - 8 | 752 | 69 | ||
| AK087134.1 | - | 7 - 8 | 807 | 69 | ||
| AK088247.1 | - | 7 - 8 | 748 | 69 | ||
| AK134406.1 | - | 7 - 8 | 722 | 69 | ||
| AK165243.1 | - | 7 - 8 | 746 | 69 | ||
| AK166118.1 | - | 7 - 8 | 746 | 69 | ||
| AK169523.1 | - | 7 - 8 | 783 | 69 | ||
| BC018191.1 | - | 7 - 8 | 732 | 3994248 | 69 | |
| BC025501.1 | - | 7 - 8 | 778 | 5321013 | 69 | |
| BC026604.1 | - | 6 - 7 | 1285 | 69 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK087134.1 transcripts.
Set Membership: |
|

Aaas
Genomic Map