Assay Details
| Gene Symbol: Atf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146065.1 | NP_666177.1 | 9 - 10 | 1006 | 120 | |
| GenBank mRNA | AK039648.1 | - | 998 | 120 | ||
| AK047901.1 | - | 982 | 120 | |||
| AK154588.1 | - | 997 | 120 | |||
| AK155046.1 | - | 9 - 10 | 997 | 120 | ||
| AK157628.1 | - | 1036 | 120 | |||
| BC026483.1 | - | 9 - 10 | 1006 | 4503463 | 120 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atf7
Genomic Map