Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500018856.mm.1 |
NM_144942.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018856.mm.1 |
NM_144942.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018856.mm.1 |
TC1500002285.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500018855.mm.1 |
NM_144942.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018855.mm.1 |
NM_144942.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018855.mm.1 |
TC1500002285.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500018858.mm.1 |
NM_144942.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018858.mm.1 |
NM_144942.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018858.mm.1 |
TC1500002285.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500018859.mm.1 |
NM_144942.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018859.mm.1 |
NM_144942.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018859.mm.1 |
TC1500002285.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500018854.mm.1 |
NM_144942.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018854.mm.1 |
NM_144942.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018854.mm.1 |
TC1500002285.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500018857.mm.1 |
NM_144942.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018857.mm.1 |
NM_144942.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500018857.mm.1 |
TC1500002285.mm.1 |
MTA TC |