Assay Details
| Gene Symbol: Rara | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177302.1 | NP_001170773.1 | 3 | 594 | 97 | |
| NM_001177303.1 | NP_001170774.1 | 2 | 339 | 97 | ||
| NM_009024.2 | NP_033050.2 | 2 | 461 | 97 | ||
| GenBank mRNA | AK042163.1 | - | 2 | 340 | 97 | |
| AK154623.1 | - | 2 | 434 | 97 | ||
| BC010216.1 | - | 2 | 459 | 3498142 | 97 | |
| BC040383.1 | - | 3 | 594 | 97 | ||
| S56656.1 | - | 2 | 441 | 97 | ||
| X56572.1 | - | 2 | 482 | 97 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK042163.1,AK154623.1,BC010216.1,NM_001177302.1,NM_001177303.1,NM_009024.2,S56656.1,X56572.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rara
Genomic Map