Assay Details
Gene Symbol: Rara | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001176528.1 | NP_001169999.1 | 4 - 5 | 1115 | 83 | |
NM_001177302.1 | NP_001170773.1 | 6 - 7 | 1320 | 83 | ||
NM_001177303.1 | NP_001170774.1 | 5 - 6 | 1065 | 83 | ||
NM_009024.2 | NP_033050.2 | 5 - 6 | 1187 | 83 | ||
GenBank mRNA | AB221572.1 | - | 4 - 5 | 84 | ||
AK042163.1 | - | 5 - 6 | 1066 | 83 | ||
AK153644.1 | - | 4 - 5 | 661 | 83 | ||
AK154623.1 | - | 5 - 6 | 1160 | 83 | ||
AK170956.1 | - | 4 - 5 | 1115 | 83 | ||
BC010216.1 | - | 5 - 6 | 1185 | 3498142 | 83 | |
BC038266.1 | - | 4 - 5 | 1184 | 83 | ||
BC040383.1 | - | 6 - 7 | 1320 | 83 | ||
CT010231.1 | - | 4 - 5 | 630 | 83 | ||
S56656.1 | - | 5 - 6 | 1167 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
